missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Genomic DNA Purification Tray I

Product Code. 15631762
Click to view available options
Quantity:
10 Trays
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 15631762

Brand: Invitrogen™ 4318641

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

TransPrep Chemistry protocol component

This 96-well Genomic DNA (gDNA) Purification Tray is used for the isolation of both genomic DNA and total RNA from homogenized plant or animal tissue samples on the ABI PRISM™ 6100 Nucleic Acid PrepSation and 6700 Automated Nucleic Acid Workstation. It is also suitable for direct DNA isolation from food and bacterial samples. The Genomic DNA (gDNA) Purification Tray is used for the TransPrep™ Chemistry protocol.
Used With DNA Elution Solution, DNA Precipitation Solution 1, DNA Precipitation Solution 2, DNA Wash Solution 1, DNA Wash Solution 2.

DNA and RNA Purification and Analysis, DNA Extraction, General gDNA Purification Reagents and Accessories, Genomic DNA Purification

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Each package contains 10 trays.
Format 96-well Plate
For Use With (Equipment) 6100 Nucleic Acid PrepStation, 6700 Automated Nucleic Acid Workstation
For Use With (Application) Nucleic Acid Purification
Quantity 10 Trays
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.