missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GeneCatcher™ gDNA Blood Kit, 0.3-1 mL

Product Code. 11550176
Change view
Click to view available options
Quantity:
96 Preps
Unit Size:
96 preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11550176 96 Preps 96 preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11550176 Supplier Invitrogen™ Supplier No. CS21101

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

GeneCatcher Magnetic Beads, Protease, Protease Buffer, GeneCatcher Lysis and Wash Buffers, and GeneCatcher Elution Buffer.

High sensitivity for archived or poorly stored blood samples

The GeneCatcher gDNA Blood Kits allow rapid and efficient extraction of genomic DNA (gDNA) from human blood. Genomic DNA is extracted from blood samples using the cost-effective, user-friendly magnetic bead based GeneCatcher™ Technology without the use of centrifugation.

  • Purity suited to highly reliable downstream processing
  • Validated in many applications, including restriction endonuclease digestion
  • Second generation multiplex and single tandem repeat (SGM+STR) profiling and quantitative PCR
  • Optimized and designed for scalable and automated gDNA isolation from large blood samples using Tecan Freedom EVO liquid handler
  • Fully scalable chemistry in batches of 8
  • High purity and yield of extracted genomic DNA yields between 300μg - 500μg per 10mL sample

DNA & RNA Purification & Analysis, DNA Extraction, Genomic DNA Purification, gDNA from Blood (Medium-Scale, 100μl-1mL)

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage The GeneCatcher™ gDNA kits include GeneCatcher™ Magnetic Beads, Protease, Protease Buffer, GeneCatcher™ Lysis and Wash Buffers, and GeneCatcher™ Elution Buffer. All components of the GeneCatcher™ gDNA Blood Kits are shipped at room temperature. Upon receipt, store the protease at +4°C and the remaining kit components at room temperature. All components are guaranteed stable for 6 months when properly stored.
Format Kit
Isolation Technology Magnetic Bead
Sample Type Blood
For Use With (Application) Real-Time Quantitative PCR (qPCR), Cloning, Southern Blotting, Sequencing, Forensic
High-throughput Compatibility Not High-throughput Compatible (Manual)
No. of Reactions 96 Preps
Prep Scale 100 μl - 1 ml Blood Samples ( Medium-Scale)
Product Line GeneCatcher
Product Type gDNA Blood Kit
Quantity 96 Preps
Shipping Condition Room Temperature
Target Genomic DNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.