missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ViraPower™ II Lentiviral Gateway™ Expression System

Product Code. 17185989
Click to view available options
Quantity:
1 kit
Unit Size:
Each
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 17185989

Brand: Invitrogen™ K36720

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Includes

6μg of pLenti6.2/V5-DEST vector (40μL vector at 150ng/μL in TE Buffer, pH 8.0), 10μg of pLenti6.2/V5-GW/lacZ plasmid (20μL of plasmid at 0.5μg/μL in TE Buffer, pH 8.0), Stbl3 Chemically Competent E. coli; 21 x 50μL of Stbl3 Cells, 6mL of S.O.C.Medium and 50μL of pUC19 control DNA (10pg/μL); ViraPower Bsd Lentiviral Support Kit includes 195μg of ViraPower Packaging Mix (Contains 195μL of a mixture of pLP1, pLP2, and pLP/VSVG plasmids, at 1μg/μL in TE Buffer, pH 8.0), 0.75mL of Lipofectamine 2000, 50mg of Blasticidin powder; 293FT Cell Line, 1 vial containing 3 X 106frozen cells in 1mL of Freezing Medium

Includes all components needed to generate lentivirus

  • Generates replication-incompetent lentivirus for transducing dividing and nondividing mammalian cells
  • The pLenti6.2/V5-DESTvector in kit has CMV promoter for driving constitutive expression of target gene in mammalian cells
  • Uses the PGK promoter instead of SV40 promoter in lentiviral vector for driving long-term, persistent expression of blasticidin stable selection marker
  • Easy recombination-based cloning and stable expression, long term expression
  • Blasticidin selection marker for stable selection under control of PGK promoter for long-term, persistent expression

Cloning, Constitutive Expression, Gateway Cloning, Mammalian Expression, Protein Expression, Proteins, Expression, Isolation and Analysis, Viral Gene Delivery

TRUSTED_SUSTAINABILITY

Specifications

Constitutive or Inducible System Constitutive
Delivery Type Lentiviral
Promoter CMV
Product Type Lentiviral Expression System
Selection Agent (Eukaryotic) Blasticidin
Content And Storage pLenti6.2⁄V5-DEST™ Gateway™ Vector Kit
• 6 μg of pLenti6.2⁄V5-DEST™ vector (40 μLofvector at 150 ng⁄μL in TE Buffer, pH 8.0)
• 10 μg of pLenti6.2⁄V5-GW⁄lacZ™ plasmid(20μL of plasmid at 0.5 μg⁄μL in TE Buffer, pH 8.0)
• One Shot™ Stbl3™ Chemically Competent E. coli(Cat # C737303): 21 x 50-μL of Stbl3™ Cells, 6 mL of S.O.C.Medium and 50 μL of pUC19 control DNA (10 pg⁄μL).

ViraPower™ Bsd Lentiviral Support Kit (Cat # K497000)
• 195 μg of ViraPower™ Packaging Mix (Contains 195μL of a mixture of pLP1, pLP2, and pLP⁄VSVG plasmids, at 1μg⁄μL in TE Buffer, pH 8.0)
• 0.75 ml of Lipofectamine™ 2000 (Cat # 11668027)
• 50 mg of Blasticidin powder (Cat # R21001)
• 293FT Cell Line (Cat # R70007): 1 vial containing 3 X 106frozen cells in 1 mL of Freezing Medium.

Storage
• Vectors: -20°C
• ViraPower™ Packaging Mix: -20°C
• Blasticidin: -20°C
• Lipofectamine™ 2000: 4°C (do not freeze)
• One Shot™ Stbl3™ Chemically Competent E. coli:-80°C
• 293 FT Cells: Liquid Nitrogen
Protein Tag V5 Epitope Tag
Cloning Method Gateway
Quantity 1 kit
Vector pLenti
Product Line Gateway, ViraPower
For Use With (Application) Viral Expression
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.