missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Gateway™ pT-Rex™-DEST31 Vector

Product Code. 11518706
Click to view available options
Quantity:
6 μg
Unit Size:
6µg
This item is not returnable. View return policy

Product Code. 11518706

Brand: Invitrogen™ 12302014

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

All destination vectors are provided lyophilized and supercoiled.

Requires

Gateway entry clone, Gateway LR Clonase™ enzyme mix, and reaction buffer.

Destination vectors for expression in E. coli, insect, yeast, or mammalian cells, as well as for production of native protein or N- or C-terminal fusion proteins

  • Destination vectors have attR sites for recombination with any attL-flanked fragment, regardless of whether it is entry clone or UltimateRF Clone

Cloning, Gateway Cloning, Mammalian Expression, Protein Expression, Proteins, Expression, Isolation and Analysis, Regulated Expression

TRUSTED_SUSTAINABILITY

Specifications

Constitutive or Inducible System Inducible
Delivery Type Transfection
Inducing Agent Tetracycline
Promoter CMV/TO
Product Type Gateway System Destination Expression Vector
Selection Agent (Eukaryotic) Geneticin™ (G-418)
Content And Storage All destination vectors are provided lyophilized and supercoiled.
Protein Tag His Tag (6x)
Cloning Method Gateway
Quantity 6 μg
Vector pDEST
Product Line Gateway, T-REx
For Use With (Application) Regulated Expression
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.