Learn More
Invitrogen™ pENTR™/TEV/D-TOPO™ Cloning Kit, with One Shot™ Mach1™-T1R Chemically Competent E. coli
Product Code. 10348192
Click to view available options
Quantity:
20 Reactions
Unit Size:
Each
For Research Use Only. All usage must comply with product instructions.
Product Code. 10348192
Brand: Invitrogen™ K253520
For Research Use Only. All usage must comply with product instructions.
Includes
pENTR/TEV/D-TOPO vector, dNTPs, salt solution, sterile water, universal M13 sequencing primers, OneShot MachI T1 Phage resistant Chemically E. coli, S.O.C. Medium, and a pUC19 control plasmid.
Use highly efficient, 5-minute cloning strategy to directionally clone blunt-end PCR product into vector
  • Blunt-end PCR products clone directionally into pENTR/TEV/D-TOPO entry vector at greater than 90% efficiency, and include TEV protease-dependent cleavage site for removal of N-terminal tags from expressed proteins
  • Rapidly shuttle cloned genes between multiple vector systems
  • Reliable performance
  • For access to Gateway System, PCR amplify gene of interest and add product straight to provided topoisomerase charged pENTR/SD/D-TOPO vector, incubate 5 minutes, and transform provided competent E. coli cells
  • Resulting attL containing Gateway Entry clones are ready for efficient recombination with choice of vectors
  • pENTR/TEV/D-TOPO vector includes Tobacco Etch Virus (TEV) recognition site for protease-dependent cleavage of an N-terminal tag from recombinant protein
  • Vector has M13 and T7 primer sequencing sites and attL recombination sites flanking PCR product insertion site, allowing clones to be easily sequence verified and recombined into choice of attR containing Gateway destination vectors
  • Kanamycin resistance gene and pUC origin are used for selection and high-copy propagation in E. coli
  • Four base overhang on vector pairs with four base sequence designed into forward primer used in PCR reaction to provide directionality to topoisomerase ligation reaction
Cloning, Gateway Cloning
TRUSTED_SUSTAINABILITY
Specifications
Cloning Method Directional TOPO™
Content And Storage pENTR™⁄TEV⁄D-TOPO™ Cloning Kit contains pENTR⁄TEV⁄D-TOPO™ vector, dNTPs, salt solution, sterile water, universal M13 sequencing primers, OneShot™ MachI™ T1 Phage resistant Chemically E. coli, S.O.C. Medium, and a pUC19 control plasmid. Store Competent E. coli at -80°C. Store all other components at -20°C.
No. of Reactions 20 reactions
Product Line One Shot
Product Type TOPO Cloning Kit
Protein Tag N-Terminal
Quantity 20 Reactions
Vector pENTR