Learn More
Invitrogen™ Gateway™ pDEST™8 Vector
Product Code. 11578666
Click to view available options
Quantity:
6 μg
Unit Size:
6µg
For Research Use Only. All usage must comply with product instructions.
Product Code. 11578666
Brand: Invitrogen™ 11804010
For Research Use Only. All usage must comply with product instructions.
Includes
All destination vectors are provided lyophilized and supercoiled.
Requires
Gateway entry clone, Gateway LR Clonase™ enzyme mix, and reaction buffer.
Destination vectors for expression in E. coli, insect, yeast, or mammalian cells, as well as for production of native protein or N- or C-terminal fusion proteins
  • Destination vectors have attR sites for recombination with any attL-flanked fragment, regardless of whether it is entry clone or UltimateRF Clone
Baculovirus Production and Titering, Cloning, Gateway Cloning, Insect Expression, Protein Expression, Proteins, Expression, Isolation and Analysis
TRUSTED_SUSTAINABILITY
Specifications
Promoter Polyhedrin
Product Type Expression Vector
Content And Storage All destination vectors are provided lyophilized and supercoiled.
Protein Tag Untagged
Cloning Method Gateway
Quantity 6 μg
Vector pDEST, Gateway Baculovirus Vectors
Product Line Gateway