Learn More
Invitrogen™ Gateway™ pDEST™15 Vector
Product Code. 11558666
Change view
Click to view available options
Quantity:
6 μg
Unit Size:
6µg
Product Code. Quantity Unit Size
11558666 6 μg 6µg
1 options
For Research Use Only. All usage must comply with product instructions.
Product Code. 11558666 Supplier Invitrogen™ Supplier No. 11802014
Black Friday Starts Here
Use promo code 27984
View all promotions
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For Research Use Only. All usage must comply with product instructions.
Includes
All destination vectors are provided lyophilized and supercoiled.
Requires
Gateway entry clone, Gateway LR Clonase™ enzyme mix, and reaction buffer.
Destination vectors for expression in E. coli, insect, yeast, or mammalian cells, as well as for production of native protein or N- or C-terminal fusion proteins
  • Destination vectors have attR sites for recombination with any attL-flanked fragment, regardless of whether it is entry clone or UltimateRF Clone
Bacterial Expression,Cloning,Gateway Cloning,Protein Expression, Proteins, Expression, Isolation and Analysis
TRUSTED_SUSTAINABILITY
Specifications
Constitutive or Inducible System Inducible
Promoter T7
Product Type Expression Vector
Selection Agent (Eukaryotic) None
Content And Storage All destination vectors are provided lyophilized and supercoiled.
Antibiotic Resistance Bacterial Ampicillin (AmpR)
Protein Tag GST Tag
Cloning Method Gateway
Quantity 6 μg
Vector pDEST, Gateway T7 Vectors
Product Line Gateway