Learn More
Invitrogen™ Baculovirus Expression System with Gateway™ Technology
Product Code. 10545003
Click to view available options
Quantity:
20 reactions
Unit Size:
Each
For Research Use Only. All usage must comply with product instructions.
Product Code. 10545003
Brand: Invitrogen™ 11827011
For Research Use Only. All usage must comply with product instructions.
Includes
LR Clonase enzyme mix and reaction buffer, destination vectors pDEST8, pDEST10, and pDEST20, proteinase K solution (2μg/μL), pENTR™-gus positive control, and Library Efficiency DH5Competent Cells. The kit does not include MAX Efficiency DH10Bac Competent Cells.
Designed to create recombinant pFastBac™ plasmids containing polyhedrin promoter
  • Destination vectors carrying polyhedrin promoter for production of native (pDEST™8), N-terminal histidine fusion (pDEST™10), and N-terminal GST fusion (pDEST™20) proteins
  • pDEST™10 vector containing TEV protease cleavage site for removal of fusion tag after protein purification
Baculovirus Production and Titering/Insect Expression/Protein Expression/Proteins, Expression, Isolation and Analysis
TRUSTED_SUSTAINABILITY
Specifications
Promoter Polyhedrin
Product Type Expression System
Content And Storage The Baculovirus Expression System with Gateway™ Technology includes LR Clonase™ enzyme mix and reaction buffer, destination vectors pDEST™8, pDEST™10, and pDEST™20, proteinase K solution (2 μg/μl), pENTR™-gus positive control, and Library Efficiency™ DH5Competent Cells. The kit does not include MAX Efficiency™ DH10Bac™ Competent Cells. Store LR Clonase™ enzyme mix and Competent Cells at -80°C. Store all other components at -20°C. Components are guaranteed stable for 6 months when properly stored.
Protein Tag GST Tag, His Tag (6x), Untagged
Cloning Method Gateway
Quantity 20 reactions
Vector pDEST, Gateway Baculovirus Vectors
Product Line Gateway