missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ExpressLink™ T4 DNA Ligase

Product Code. 11370862
Click to view available options
Quantity:
30 Reactions
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11370862

Brand: Invitrogen A13726

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

ExpressLink™ T4 DNA Ligase and 5X reaction buffer [250mM Tris-HCl (pH 7.6), 50mM MgCl2, 5mM ATP, 5mM DTT, 25% (w/v) polyethylene glycol-8000].

Catalyzes the ligation of blunt- or cohesive-end DNA fragments in only 5 minutes at room temperature (25°C), and PCR fragments with A overhangs in 15 minutes at 25°C

Single-stranded nucleic acids are not substrates for this enzyme.

T4 DNA ligase catalyzes the formation of phosphodiester bonds between double-stranded DNA strands with 3' hydroxyl and 5' phosphate termini in the presence of ATP. ExpressLink™ T4 DNA Ligase formulation is optimized for faster reaction times and more convenient incubation temperature than our other T4 DNA Ligase formulations.

See the Difference the Cleanest Ligase Makes
We apply the most stringent test available for exonuclease contamination to every lot of ExpressLink™ T4 DNA Ligase. The superior sensitivity of our assay will detect much lower active exonuclease concentrations than assays used by other manufacturers. As a result, we offer one of the best ligase products on the market. You can be confident of achieving the highest quality data in your important experiments with ExpressLink™ T4 DNA Ligase.

Performance and Quality Testing: Endodeoxyribonuclease, 3' and 5' exodeoxyribonuclease assays; ligation efficiency tested.

Unit Definition: One unit catalyzes the exchange of 1nmol 32P-labeled pyrophosphate into ATP in 20 min. at 37°C (One unit is equal to approximately 300 cohesive-end ligation units).

Unit Reaction Conditions: 66mM Tris-HCl (pH 7.6), 6.6mM MgCl2, 10mM DTT, 66μM ATP, 3.3μM 32P-labeled pyrophosphate, and ExpressLink™ T4 DNA Ligase in 0.1mL for 20 min. at 37°C.

  • Fast, 5 minute ligation of blunt- or cohesive-end DNA fragments
  • Ligation of PCR fragments with “A” overhangs in 15 minutes
  • Convenient 25°C reaction temperature eliminates the need for a water bath or temperature block
  • Ideal for cloning DNA into vectors, recircularization of linear DNA, library construction, TA cloning, and linker ligation
  • Superior exonuclease quality control assay for exonuclease-free ligations

ExpressLink™ T4 DNA Ligase is unstable when kept on ice for long periods. It is recommended that the enzyme be kept at –20°C until within 5–10 minutes of use and returned immediately to –20°C after use.

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Concentration 5 U/μL
Components ExpressLink™ T4 DNA Ligase and 5X reaction buffer [250mM Tris-HCl (pH 7.6), 50mM MgCl2, 5mM ATP, 5mM DTT, 25% (wG/v) polyethylene glycol-8000].
Content And Storage -20°C
Source E. coli expressing a T4 DNA ligase gene.
Quantity 30 Reactions

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.