missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ElectroMAX™ DH10B T1 Phage-Resistant Competent Cells

Product Code. 12007519
Change view
Click to view available options
Quantity:
5 x 100 μL
Unit Size:
0.5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12007519 5 x 100 μL 0.5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12007519 Supplier Invitrogen™ Supplier No. 12033015
  • ONLINE EXCLUSIVE PRICE
    £392.00
    £380.65 / 0.5mL
    Save £11.35
    2% Off
View all promotions
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

ElectroMAX DH10B T1 Phage-Resistant Cells: 5 vials, 100μL each, pUC19 DNA (10pg/μL): 1 vial, 50μL, S.O.C. medium: 2 bottles, 6mL each

ElectroMAX DH10B T1 Phage-Resistant Competent Cells offer transformation efficiencies that are among the highest we offer and can exceed 1 x 1010 cfu/μg plasmid DNA in efficiency.

ElectroMAX DH10B T1 Phage-Resistant Competent Cells offer transformation efficiencies that are among the highest we offer and can exceed 1 x 1010 cfu/μg plasmid DNA in efficiency. They are ideal for applications requiring high-efficiency transformation, including cDNA and gDNA library construction.

ElectroMAX DH10B cells:
• Maximize yield of transformants from limited cloning products
• Permit efficient cloning of methylated DNA
• Are resistant to T1 and T5 bacteriophages
• Support blue/white screening by α-complementation on plates containing X-Gal or Bluo-Gal
• Deliver high-yield plasmid preparations for downstream applications

Phage-resistant representative libraries from a single transformation
The tonA mutation in DH10B cells confers resistance to bacteriophages T1 and T5, safeguarding valuable libraries against contamination. Additionally, mutations in the methylation-dependent restriction system (mcrA, mcrBC, and mrr) make ElectroMAX DH10B T1 Phage-Resistant Cells ideal for construction of genomic libraries of both prokaryotic and eukaryotic genomic DNA and allow for efficient plasmid rescue from eukaryotic genomes. ElectroMAX DH10B T1 Phage-Resistant Cells are suitable for construction of gene banks, for the generation of cDNA libraries using plasmid-derived vectors, and for situations when DNA is limiting. This strain also has the Φ80lacZΔM15 genotype, providing for the option of blue/white screening on plates containing either X-Gal or Bluo-Gal. Finally, the endA1 mutation makes DH10B an excellent vehicle for amplifying plasmid DNA for subsequent extraction and purification.

Genotype
F-mcrA Δ(mrr-hsdRMS-mcrBC) Φ80lacZΔM15 ΔlacX74 recA1 endA1 araD139Δ(ara, leu)7697 galU galK λ-rpsL nupG tonA

Find the strain and format that fits your needs
• DH10B cells are available in both electrocompetent and chemically competent formats.
• ElectroMAX DH10B cells (Cat. No.18290015) are suitable for cDNA or gDNA library construction but are not resistant to T1 and T5 phage infection.
• MegaX DH10B T1R Electrocomp Cells (Cat. No. C640003) have the highest transformation efficiency (>3 x 1010 cfu/μg plasmid DNA) and also offer the benefit of T1 and T5 phage resistance.

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Product Type Electrocompetent Cells
Contains F' Episome No
Improves Plasmid Quality Yes (endA1)
Cloning Methylated DNA Yes (mcrA)
Transformation Efficiency Level High Efficiency (>1 x 109 cfu/μg)
Content And Storage • ElectroMAX DH10B T1 Phage-Resistant Cells (5 x 100 μL)
Store Competent Cells at –80°C.

• pUC19 DNA (50 μL at 10 pg/μL)
Store pUC19 DNA at –20°C.

• S.O.C. Medium (2 x 6 mL)
Store S.O.C. Medium at 4°C or room temperature.

.
Antibiotic Resistance Bacterial Yes (Streptomycin)
Cloning Unstable DNA Not suitable for cloning unstable DNA
Blue/White Screening Yes (lacZΔM15)
High-throughput Compatibility Low
Plasmid May be used for plasmids >20 kb
Preparing Unmethylated DNA No
Reduces Recombination Yes (recA1)
Shipping Condition Dry Ice
T1 Phage - Resistant (tonA) Yes
Species E. coli (K12)
Format Tube
Product Line ElectroMAX
Quantity 5 x 100 μL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.