missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ E-Gel™ High-ReSolution Agarose Gels, 4%
Description
Includes
Contains one multi-pack of 18 gels.
- Ready for use with agarose, electrodes, and DNA stain packaged inside a dry, disposable, UV-transparent cassette
- There are no gels to pour, buffers to make, staining or destaining steps to perform, or gel boxes to assemble
- Offer excellent resolution and clarity in 12 minutes and are excellent for analyzing PCR products, restriction digests, and plasmid preparations
- 12-well, single comb gels
Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Content And Storage | Contains one multi-pack of 18 gels. Store at room temperature. |
| Gel Percentage | 4% |
| Gel Type | E-Gel |
| Wells | 12-well |
| In Gel Dye | Ethidium Bromide |
| Shipping Condition | Room Temperature |
| Well Design | Single Comb |
| Product Line | E-Gel |
| Product Type | Agarose Gel |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?