missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ E-Gel™ Go! Agarose Gels, 2%

For performing agarose gel electrophoresis with only a few samples to analyze

Brand:  Invitrogen™ G442002

Code : 50

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12013589

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • Precast agarose gel cassettes each have four wells and are premixed with a blue light excitable nucleic acid stain
  • Can be stored at room temperature and are ready to use without additional buffers or reagents
  • Designed specifically to work with the components of the E-Gel Go! System
  • For a quick check of end point PCRs, restriction digests, plasmid preps or other molecular biology reactions
  • Use proprietary stain formulated to provide the high detection sensitivity and can be visualized with traditional UV transilluminators
  • When using blue light the bands must be viewed through an amber filter
  • Sharp bands and repeatable, reliable results
  • Single comb well format
  • Sample Volume; 10μL

  • For linear double stranded DNA fragments ranging from 1.5kbp to 100bp in size the 2% gels offer excellent resolution and separation

Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting

Specifications

Specifications

E-Gel™ 1 Kb Plus Express Ladder
2%
Go Agarose Gel
50 to 2000 bp
4-well
Room Temperature
Single Comb
• Each pack contains 20 individually sealed 2% E-Gel™ Go! precast agarose gel cassettes

Store at room temperature.
E-Gel
10 μL
SYBR™ Gold
20 Gels/Pk.
E-Gel™ Go!
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.