missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ E-Gel™ General Purpose Agarose Gels, 2%

Product Code. 12060136
Change view
Click to view available options
18 gels
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
12060136 18 gels
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12060136 Supplier Invitrogen™ Supplier No. G501802

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

Contains one multi-pack of 18 gels.

Self-contained, precast agarose gels designed to provide fast, convenient, and easy electrophoresis

  • Ready for use with agarose, electrodes, and DNA stain packaged inside a dry, disposable, UV-transparent cassette
  • There are no gels to pour, buffers to make, staining or destaining steps to perform, or gel boxes to assemble
  • Offer excellent resolution and clarity in 12 minutes and are excellent for analyzing PCR products, restriction digests, and plasmid preparations
  • 12-well, single comb gels

Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains one multi-pack of 18 gels. Store at room temperature.
Gel Percentage 2%
Gel Type E-Gel
Wells 12-well
In Gel Dye Ethidium Bromide
Shipping Condition Room Temperature
Well Design Single Comb
Product Line E-Gel
Product Type Agarose Gel
compliance-icons
Product Identifier
  • E-Gel 2% General Purpose Agarose 18-Pak
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 3(2H)-isothiazolone,5-chloro-2-methyl-with 2-methyl-3(2H)-isothiazolone

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.