missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ E-Gel™ General Purpose Agarose Gels, 0.8%

Product Code. 12070136
Click to view available options
:
18 gels
This item is not returnable. View return policy

Product Code. 12070136

Brand: Invitrogen™ G501808

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

Contains one multi-pack of 18 gels.

Self-contained, precast agarose gels designed to provide fast, convenient, and easy electrophoresis

  • Ready for use with agarose, electrodes, and DNA stain packaged inside a dry, disposable, UV-transparent cassette
  • There are no gels to pour, buffers to make, staining or destaining steps to perform, or gel boxes to assemble
  • Offer excellent resolution and clarity in 12 minutes and are excellent for analyzing PCR products, restriction digests, and plasmid preparations
  • 12-well, single comb gels

Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains one multi-pack of 18 gels. Store at room temperature.
Gel Percentage 0.8%
Gel Type E-Gel
Product Type Agarose Gel
Wells 12-well
In Gel Dye Ethidium Bromide
Product Line E-Gel
Shipping Condition Room Temperature
Well Design Single Comb

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.