missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ E-Gel™ EX Agarose Gels Starter Kit, 1%
Click to view available options
Quantity:
Each
Unit Size:
Each
Description
Includes
The E-Gel EX Starter Kits include the E-Gel iBase Power System, the E-Gel Safe Imager Real-Time Transilluminator, E-Gel 1Kb Plus DNA Ladder, and a pack of 10 gels containing either 1% or 2% E-Gel EX gels.
- Complete separation (by molecular weight) in 10 minutes
- Easy access to the gel for extraction of DNA bands or transfer to membranes for Southern blotting DNA Separation and sensitivity
- Demonstrate over five-fold greater sensitivity than comparable gels containing ethidium bromide
- Superior sensitivity requires less sample, saving time and money
- Can also be used for fast analysis of RNA samples to check on their integrity before proceeding with downstream applications
- A proprietary stain incorporated in the gels is excitable by visible blue light, allowing safe, real-time monitoring of DNA or RNA migration with the E-Gel Safe Imager Real-Time Transilluminator
- The room-temperature stable, ready-to-use 1Kb Plus DNA Ladder is premixed with loading buffer and is formulated specifically for E-Gel EX gels as well as other E-Gel precast gels
- Standard ladders not premixed with loading buffer may also be used, but in significantly lower amounts than required for other gels
- Single Comb
- Sample Type: Double-Stranded DNA (dsDNA), Single-Stranded RNA
- Sample Volume; 20μL
Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting
Specifications
Specifications
| Concentration | 1% |
| Format | Multiwell |
| Product Type | Agarose Gels Starter Kit |
| Separation Range | 100 to 5000 bp |
| Wells | 11-well |
| Kit Contents | The E-Gel EX Starter Kits include the E-Gel iBase Power System, the E-Gel Safe Imager Real-Time Transilluminator, E-Gel 1Kb Plus DNA Ladder, and a pack of 10 gels containing either 1% or 2% E-Gel EX gels. Packs of 20 gels and 10 gels are also available separately. |
| Quantity | Each |
| Storage Requirements | Store gels at room temperature. E-Gel™ EX gels are guaranteed stable for 3 months when properly stored. |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction