Learn More
Invitrogen™ E-Gel™ EX Agarose Gels, 1%
Precast, fast-resolving gels with 11 wells and an openable format
Brand: Invitrogen™ G402021
240.65 GBP valid until 2024-12-31
Use promo code "22910" to get your promotional price.
Additional Details : Weight : 1.12000kg
Requires : Require the E-Gel iBase™ Power System for running and the E-Gel Safe Imager™ Real-Time Transilluminator for real-time viewing of sample migration.
Description
- Contain a proprietary fluorescent nucleic acid stain (excitation: 490nm, emission: 520nm) that can be viewed with blue light, which minimizes DNA damage and allows detection down to 1ng/band of DNA
- Complete separation (by molecular weight) in 10 minutes
- Live monitoring of DNA migration without the hazards of UV light
- Easy access to the gel for extraction of DNA bands or transfer to membranes for Southern blotting
- Demonstrate over five-fold greater sensitivity than comparable gels containing ethidium bromide allows less sample use per run
- Fast analysis of RNA samples to check on their integrity before proceeding with downstream application
- The gel cassette is designed to open with a Gel Knife, so that the gel inside can be accessed readily for excision of specific bands or transferred to a membrane for Southern blot analysis
- Single comb well format
- Sample Type: Double-Stranded DNA (dsDNA), Single-Stranded RNA
- Sample Volume; 20μL
Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting
Order Info
Shipping Condition: Room Temperature
Specifications
E-Gel™ 1 Kb Plus Express Ladder, Millennium™ RNA Markers | |
1% | |
EX Agarose Gel | |
100 to 5000 bp | |
11-well | |
20 Gels/Pk. | |
Double-Stranded DNA (dsDNA), Single-Stranded RNA, Circular RNA | |
Single Comb |
• E-Gel™ EX 20-Paks contain 20 gels Store gels at room temperature. Guaranteed stable for 3 months when properly stored. |
|
E-Gel | |
20 μL | |
SYBR™ Gold | |
Molecular Weight | |
Preparative without dyes, Non-preparative | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.