Learn More
Invitrogen™ E-Gel™ CloneWell™ Agarose Gels with SYBR Safe™ DNA Gel Stain, 0.8%
Less hazardous
Brand: Invitrogen™ G661808
Code : 50
Additional Details : CAS Number : 79-06-1 Weight : 0.00450kg
Includes: Contains 18 gels
Description
- For isolating DNA bands separated by agarose gel electrophoresis
- No additional gel purification steps required after electrophoresis
- Remove purified DNA directly from the well with a pipette
- Minimize DNA damage and get more colony forming units
- DNA extracted from CloneWell gels is compatible with common cloning methods such as restriction cloning, TOPO™ cloning, and Gateway™ cloning
- Features two rows of wells: To extract DNA band of interest without conventional gel purification, load sample into the top row and electrophorese until the band moves into the bottom row of wells; Then use a pipette to remove purified DNA band
- Guides on the gel cassette help track the band of interest
- The 0.8% agarose concentration is optimal for separating bands up to 5 kb in length
- SYBR™ Safe DNA Gel Stain is incorporated into the gel itself, allowing the user to visually monitor migration on a blue-light transilluminator
Agarose Gel Electrophoresis, Agarose Gel Extraction, DNA and RNA Purification and Analysis, DNA Extraction, Nucleic Acid Gel Electrophoresis and Blotting, PCR Product Clean-Up
Order Info
Shipping Condition: Room Temperature
Specifications
0.8% | |
Contains 18 gels | |
Room Temperature |
Agarose Gel Cassette | |
18 Pack |
For Research Use Only. Not for use in diagnostic procedures.