missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ E-Gel™ 96 High Range DNA Marker

Product Code. 10338412
Change view
Click to view available options
Quantity:
100 applications
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10338412 100 applications Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10338412 Supplier Invitrogen™ Supplier No. 12352019
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

The E-Gel™ High Range DNAmarker is supplied at 10μg/mL in 10mM Tris-HCl (pH 7.5), 1mM EDTA, 0.0025% bromophenol blue, 5% glycerol; For 100 applications

Suitable for sizing doublestranded DNA on all E-Gel™ gel formats

Designed specifically for clear band separation on E-Gel™ 96 gels, this ladder contains 5 bands ranging in size from 400 to 10,000bp. This ladder is supplied premixed in an optimized E-Gel loading buffer. Compatible with E-Gel™, E-Gel™ 48, E-Gel™ 96 gels.

Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting

Order Info

Shipping Condition: Approved for shipment at Room Temperature or on Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 2 x 1 mL E-Gel 96 High Range DNA Ladder
• 1 mL 1X E-Gel Sample Loading Buffer

Store at room temperature or at 4°C for up to 6 months.
For longer periods, store at -20°C.
Concentration 0.005 μg/μL
Gel Type Agarose
Ready to Load Yes
Sample Loading Volume 1 mL
Size Range 400 to 10000 bp
Volume (Metric) 2 x 1000 μL
Gel Compatibility E-Gel™ agarose gels
Quantity 100 applications
Shipping Condition Approved for shipment at Room Temperature or on Dry Ice
Technology Individual chromatography-purified DNA fragments
Product Type High Range DNA Marker
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.