missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ Untouched™ Mouse CD4 Cells Kit

Product Code. 10252133
Change view
Click to view available options
Quantity:
4 mL
2 x 10 mL
Unit Size:
4mL
Pack of 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10252133 2 x 10 mL Pack of 2
10236794 4 mL 4mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10252133 Supplier Invitrogen™ Supplier No. 11415D

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

20mL Mouse Depletion Dynabeads and 2mL Antibody Mix with monoclonal antibodies towards CD8, CD11b, CD16/32, CD45R and Ter-119. These reagents are sufficient to process approx. 1 x 109 cells

Dynabeads™ Untouched™ Mouse CD4 Cells contains both magnetic beads and an Antibody Mix to deplete all leucocytes from mouse spleen or lymph node cells, leaving only the target mouse CD4+ cells.

Dynabeads Untouched Mouse CD4 Cells supplies both 4.5μm superparamagnetic Dynabeads (with a polyclonal sheep anti-rat IgG antibody covalently bound to the bead surface) and a monoclonal Antibody Mix, together designed for the depletion of CD4-negative leucocytes from mouse spleen or lymph node single-cell suspensions. Following the depletion procedure, the negatively isolated, untouched target mouse CD4+ cells are pure, viable, and ready to be used in any downstream application.

  • Rapid isolation of untouched mouse CD4
  • T cells—no columns required
  • Pure and viable mouse CD4+ T cells are ready for any downstream application

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Cell Type T Cells (CD4)
Content And Storage Contains: 20 mL Mouse Depletion Dynabeads™ and 2 mL Antibody Mix with monoclonal antibodies towards CD8, CD11b, CD16/32, CD45R and Ter-119. These reagents are sufficient to process ∼1 x 109 cells.

Store kit components at 2°C to 8°C.
Product Type Cell Isolation Kit
Isolation Technology Negative Isolation
No. of Cells Processes ∼1 x 109 cells total
Output Viability >95%
Product Line DYNAL, Dynabeads
Purity or Quality Grade Research Grade
Quantity 2 x 10 mL
Reactivity Mouse
Sample Type Lymph Node, Spleen
Shipping Condition Room Temperature
Starting Material Cell No. 1 x 107 cells per isolation
Target Species Mouse
Diameter (Metric) 4.5 μm
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.