Learn More
Invitrogen™ Dynabeads™ Untouched™ Human Monocytes Kit
Description
Includes
10mL Depletion MyOne; SA Dynabeads and 2mL Antibody Mix with biotinylated monoclonal antibodies towards human CD3, CD7, CD16 (a and b), CD19, CD56, CD123 and CD235a (Glycophorin A)
The Dynabeads Untouched Human Monocytes Kit is used to isolate pure and viable monocytes from PBMC by negative isolation. The kit depletes T cells, B cells, NK cells, dendritic cells, granulocytes, and erythrocytes, while the negatively isolated human monocytes are left in the sample. The isolated monocytes cells are in perfect shape for any functional assay and application. Advantages of the Dynabeads Untouched Human Monocytes Kit:
• High purity, recovery, and viability of human monocytes
• Easy to use and to scale up
• No columns required
Magnetic bead-based separation offers easy handling
An antibody mix towards the non-monocytes is added to the sample and allowed to bind to the cells. After a quick wash, the Dynabeads are added and will bind to the antibody-labeled cells during a short incubation. The bead-bound cells are quickly separated on a magnet and discarded. The remaining negatively isolated human monocytes can be directly analyzed in a flow cytometer and used in any application.
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Cell Type | Monocytes |
| Content And Storage | The kit contains: 10 mL Depletion MyOne SA Dynabeads, 2 mL Blocking Reagent, and 2 mL Antibody Mix with biotinylated monoclonal antibodies towards human CD3, CD7, CD16 (a and b), CD19, CD56, CD123 and CD235a (glycophorin A) Storage conditions: 2–8°C. Do not freeze. |
| Product Type | Human Monocytes Kit |
| Final Product Type | Cells |
| Format | Bead |
| Isolation Technology | Negative Isolation |
| No. of Cells | Processes ∼1 x 109 cells/kit |
| No. of Tests | 20 |
| Output Viability | >96% |
| Product Line | DYNAL, Dynabeads, Untouched |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.