Learn More
Invitrogen™ DYNAL™ Dynabeads™ Sequencing Clean-Up Kit
Offers a simple one-tube system for purifying DNA sequencing reactions
£156.00 - £784.00
Specifications
Content And Storage | 2°C to 8°C |
---|---|
For Use With (Application) | DNA and RNA Purification and Analysis, DNA Extraction, Sequencing Reaction Purification |
Includes | Dynabeads in binding buffer |
Sample Type | Sequencing Reactions |
Includes: Dynabeads in binding buffer
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10242683
|
Invitrogen™
66102 |
5 mL |
£784.00
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
10592045
|
Invitrogen™
66101 |
1 mL |
£156.00
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||||
Description
Removal of unincorporated dye terminators, salts and other unwanted components from extension products is critical for both standard and capillary DNA sequencing systems. The unique uniformity of beads guarantees consistent, high quality results, long reading lengths and high Phred 20 scores.
- Efficient removal of excess dye terminators
- Rapid and simple, one-tube protocol can be completed in 15 minutes
- No centrifugation or filtration
- Generate sequences with high number of Phred 20 bases
- Effective elimination of dye-blobs
- High throughput compatible
- Purification Target: DNA from Enzymatic Reactions (e.g. PCR), Sequencing Reactions
- Sample Type: Sequencing Reactions
Please note that this product is not for sale or use in the USA.
DNA and RNA Purification and Analysis, DNA Extraction, Sequencing Reaction Purification
Order Info
Shipping Condition: Room Temperature
Specifications
2°C to 8°C | |
DNA and RNA Purification and Analysis, DNA Extraction, Sequencing Reaction Purification | |
Dynabeads in binding buffer | |
Sequencing Reactions |
For Research Use Only. Not for use in diagnostic procedures.