missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ M-280 Streptavidin

For solation and handling of biotinylated nucleic acids, antibodies, or other biotinylated ligands and targets

Brand:  Invitrogen™ 60210

Product Code. 15534364

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Dynabeads are used on more than 25,000 routine IVD instruments worldwide and are known for exceptional reproducibility (both within and between batches) and automation ability. The strong binding affinity of streptavidin-biotin interaction (Kd=10-15) is used in vast number of applications.

  • Direct and fast isolation of any biotinylated molecule
  • Low-charged and neutral beads, optimal for binding of DNA fragments, proteins, peptides, and antibodies
  • Flexible protocols with gentle and efficient liquid-phase reaction kinetics
  • Biomagnetic protocols are easily adapted to automated platforms
  • High batch-to-batch reproducibility, securingconsistent results in application
  • Bead Diameter: 2.8 µm
  • Concentration: 10mg/mL
  • High throughput compatible
  • Sample Type: Any Sample Type
  • Diameter: 2.8°m, with monolayer, not a multilayer, of recombinant streptavidin covalently coupled to surface and further blocked with BSA
  • Show rapid liquid-phase reaction kinetics.
  • Key applications: Preparing 2 to 5kb samples for mate pair library sequencing, isolation of RNA and DNA binding proteins, immobilization of large DNA fragments, purifying sequencing products, specific capture of nucleic acids
  • Dynabeads™ are used on more than 25,000 routine IVD instruments worldwide
One mg of Dynabeads M-280 Streptavidin typically binds:
  • 650 to 900 pmoles free biotin
  • ∽200 pmol biotinylated peptides
  • ∽10µg biotinylated IgG
  • ∽10µg ds-DNA
  • ∽200 pmol ss-oligonucleotides

Antibody Purification, ChIP-on-Chip, Chromatin Biology, Chromatin Immunoprecipitation (ChIP), DNA and RNA Purification and Analysis, DNA Extraction, Immunoprecipitation, Phage Display, Protein and Peptide Purification, Protein Complex Isolation and Analysis, Protein Complex Pull-Down, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis, RNAi, Epigenetics and Non-Coding RNA Research, Sanger Sequencing, Sequencing, Sequencing Reaction Purification

TRUSTED_SUSTAINABILITY
Specifications

Specifications

650-900 pmoles/mg beads
This Product Contains: Dynabeads™ (2.8 µm) with covalently coupled recombinant streptavidin. Supplied in PBS, pH 7.4 with 0.1% BSA and 0.02% sodium azide.

Store at: 2°C to 8°C.
Beads in Suspension
Streptavidin
Any Sample Type
Tosylactivated, Hydrophobic
10 mg⁄ml, 10 mg/mL
Streptavidin
High-throughput Compatible
DYNAL™, Dynabeads™
Room Temperature
2.8 μm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.