missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ DNA DIRECT™ Universal Kit

Product Code. 10785403
Change view
Click to view available options
Quantity:
300 Preps
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10785403 300 Preps Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
To receive the discount customers must purchase 2 of the same product at list price in a single order to receive 30% discount. There is no limit to the multiples of 2 that customers can buy. Use promo code ”27974” to get your promotional price
Product Code. 10785403 Supplier Invitrogen™ Supplier No. 63006

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
To receive the discount customers must purchase 2 of the same product at list price in a single order to receive 30% discount. There is no limit to the multiples of 2 that customers can buy. Use promo code ”27974” to get your promotional price

Includes

Dynabeads and all neccesary buffers and solutions

Contaminating PCR inhibitors are eliminated without any centrifugation steps or use of phenol/chloroform.

Contaminating PCR inhibitors are eliminated without any centrifugation steps or use of phenol/chloroform. The kit is particularly useful for isolation of DNA from bacteria and cultured cells, as well as from clinical specimens and tissues from various species.

  • User-friendly 10 minute isolation of genomic DNA
  • Only minute amount of starting material is required, even finger-prick samples can be used
  • Protocols have been developed for fully automated use on liquid-handling robots, e.g. on Biomek; 2000, from Beckman Coulter, Genesis RSP from Tecan, etc
  • Isolation of PCR-ready genomic DNA directly from crude material

Not high-throughput compatible (Manual)

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in refrigerator (2–8°C).
Isolation Technology Magnetic Bead
Sample Type Fungi, Bacteria, Tissue, Blood, Cells, Buccal Samples
For Use With (Application) Sequencing
High-throughput Compatibility Not High-throughput Compatible (Manual)
Product Line DNA DIRECT, DYNAL, Dynabeads
Product Type Universal Kit
Quantity 300 Preps
Shipping Condition Room Temperature
Target Genomic DNA

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.