missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ CD45

Product Code. 15414693
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
This item is not returnable. View return policy

Product Code. 15414693

Brand: Invitrogen™ 11153D

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

5mL Dynabeads CD45 coated with anti-CD45 monoclonal antibody.

Dynabeads™ CD45 are superparamagnetic beads covalently coupled to anti-human CD45 antibody that enable isolation or depletion of CD45+ leucocytes directly from whole blood, buffy coat, or MNC suspensions.

Dynabeads™ CD45 are superparamagnetic beads covalently coupled to anti-human CD45 antibody that enable isolation or depletion of CD45+ leucocytes directly from whole blood, buffy coat, or MNC suspensions. Dynabeads™ CD45 can also be used to enrich epithelial tumor cells. The CD45 antigen is weakly expressed on myeloid cells. To efficiently deplete or isolate all leucocytes including myeloid cells, use Dynabeads™ CD15 (Cat. No. 11137D) in combination with Dynabeads™ CD45. Advantages of Dynabeads™ CD45:

• Efficient depletion of human leucocytes
• Isolation of leucocytes directly from whole blood for molecular applications
• Enrichment of circulating epithelial tumor cells

About Dynabeads™ CD45
Dynabeads™ CD45 are uniform, superparamagnetic beads (4.5 μm diameter) coated with a primary monoclonal mouse IgG2a antibody specific for a CD45 membrane antigen common to all known isoforms of CD45. CD45 is expressed on all human leucocytes. Because of the bead size, Dynabeads™ CD45 can easily and efficiently isolate or deplete cells from viscous samples such as whole blood and bone marrow in about 30 minutes. Dynabeads™ CD45 is often used for enrichment of circulating non-hematopoietic tumor cells, by depletion of all CD45+ leucecytes from a MNC sample. Positively isolated cells can be used for downstream molecular studies; for example, those in which cells are to be lysed while still attached to beads, and nucleic acids or proteins further purified. Note that intact cells will not be released from these beads, and we do not recommend to analyze the bead-bound cells with a flow cytometer.

Magnetic bead–based separation offers easy handling
Dynabeads™ CD45 is added to the sample under continuous mixing to optimize the binding of the Dynabeads™ to the target cells. By placing the sample on a magnet it separates the bead-bound target cells from the rest of the sample in just 1–2 minutes. For depletion, remove the supernatant to a new tube for further studies and discard the bead-bound cells. For positive isolation for molecular studies, remove the supernatant and wash the bead-bound cells 2–3 times in buffer to get optimal purity. The cells can be lysed while still attached to the beads and the supernatant transferred to a new tube for downstream molecular analysis. Starting samples can be whole blood, buffy coat, bone marrow, PBMC, or tissue digests.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Cell Type Tumor Cells, Leukocytes
Content And Storage This Product Contains: 5 mL Dynabeads™ CD45 coated with anti-CD45 monoclonal antibody.

Storage Conditions: 2°C to 8°C. Do not freeze.
Product Type Antibody Coated Bead
Isolation Technology Depletion, positive isolation for molecular applications
No. of Cells Processes ∼5 x 108 cells total
Output Viability >95%
Product Line DYNAL, Dynabeads
Purity or Quality Grade Research Grade
Quantity 5 mL
Sample Type PBMC, Blood
Shipping Condition Room Temperature
Starting Material Cell No. 1 x 107 PBMCs per isolation
Target Species Human
Diameter (Metric) 4.5 μm
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.