missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ CD25

Product Code. 10690644
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10690644 5 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10690644 Supplier Invitrogen™ Supplier No. 11157D

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

5mL Dynabeads CD25 coated with anti-CD25 monoclonal antibody

Isolate CD25+ T-cells directly from whole blood or PBMC. Dynabeads™ CD25 are ideal for depleting CD25+ T-cells and for direct positive isolation for downstream molecular analysis.

Isolate CD25+ T-cells directly from whole blood or PBMC. Dynabeads™ CD25 are ideal for depleting CD25+ T-cells and for direct positive isolation for downstream molecular analysis.

• Depletion of CD25+ T-cells
• Isolation of pure and viable CD25+ T-cells for molecular applications
• No columns required

Note: For isolation of human CD4+CD25+Foxp3+ regulatory T-cells (Treg cells), please use Dynabeads™ Regulatory CD4+CD25+ T Cells (Cat. no. 113-63D).

Starting samples:
Whole blood, PBMC, buffy coat.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Cell Type T Cells (CD25)
Content And Storage This Product Contains: 5 mL Dynabeads™ CD25 coated with anti-CD25 monoclonal antibody.

Store at 2°C to 8°C.
Product Type Antibody Coated Bead
Isolation Technology Depletion, positive isolation for molecular applications
No. of Cells Processes ∼5 x 109 cells total
Output Viability >95%
Product Line DYNAL, Dynabeads
Purity or Quality Grade Research Grade
Quantity 5 mL
Sample Type PBMC, Blood
Shipping Condition Room Temperature
Starting Material Cell No. 2.5 x 107 PBMCs per isolation
Target Species Human
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.