missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ M-270 Amine

Product Code. p-4924177
Click to view available options
Quantity:
2 mL
10 mL
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10151003

Brand: Invitrogen™ 14307D

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

These are hydrophilic Dynabeads™ (2.8 μm) with amine groups. The surface amino groups allow direct covalent binding of carbohydrates, glycoproteins and glycolipids through reductive amination of aldehydes.

The surface amino groups allow direct covalent binding of carbohydrates, glycoproteins and glycolipids through reductive amination of aldehydes. For use with any sample type.

  • Rapid binding (less than 1 hour) at neutral to high pH and room temperature
  • No further surface activation required
  • Excellent reactivity with a range of cross-linking reagents, allowing for easy introduction of further alternative surface chemistries
  • Ideal for isolation of small components such as proteins and peptides
  • Allows C-terminal coupling of proteins
  • Well suited for isolation of proteins or other targets with affinity for specific carbohydrate moieties

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in refrigerator (2–8°C).
Diameter (Metric) 2.8 μm
Elution Volume Min 5 μL
High-throughput Compatibility High-throughput Compatible
Shipping Condition Room Temperature
Sample Type Any Sample Type
Surface Functionality Amine
Product Line Dynabeads
Quantity 2 mL
Type Bead with Amine Group
Format Beads in suspension
Purity or Quality Grade Research Grade
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.