missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DNAzol™ BD Reagent, for isolation of genomic DNA from whole blood

Product Code. 11518646
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11518646 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 11518646 Supplier Invitrogen™ Supplier No. 10974020

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

DNAzol BD Reagent is a ready-to-use organic reagent specifically formulated to react in about 30 minutes.

DNAzol BD Reagent is a ready-to-use organic reagent specifically formulated to react in about 30 minutes.

  • Specifically formulated for extraction of genomic DNA from whole blood
  • Optimized for rapid Isolation of genomic DNA
  • Efficient isolation of genomic DNA from whole blood
  • Rapid isolation of genomic DNA
  • Isolated DNA used for many downstream applications

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
Isolation Technology Organic Extraction
Sample Type Blood
Final Product Type Genomic DNA
For Use With (Application) PCR, Southern Blotting, Sequencing, Cloning
High-throughput Compatibility Not High-throughput Compatible (Manual)
Product Line DNAzol
Product Type BD Reagent
Quantity 100 mL
Shipping Condition Room Temperature
Target Genomic DNA
Test Time 30 min.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.