missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ Regulatory CD4+/CD25+ T Cell Kit

Product Code. 10154833
Change view
Click to view available options
Quantity:
2 x 5 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10154833 2 x 5 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10154833 Supplier Invitrogen™ Supplier No. 11363D

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

2mL Antibody Mix Human CD4, 10mL Depletion MyOne; Dynabeads, 5mL Dynabeads CD25 and 2mL DETACHaBEAD™. These reagents are sufficient to process approx. 1 x 109 cells

Use this kit to isolate highly pure CD4+ CD25+ regulatory T-cells (Treg) that express the intracellular transcription factor Foxp3.

Use this kit to isolate highly pure CD4+ CD25+ regulatory T-cells (Treg) that express the intracellular transcription factor Foxp3. The CD4+ CD25- cell fraction can be used as effector cells in downstream inhibitory assays. You can expect >95% purity (CD4+ CD25+ expression) and >80% Foxp3 expression.

Isolate bead- and antibody-free human CD4+ CD25+ Foxp3+ Treg cells
• Treg cells can be expanded with Dynabeads™ CD3/CD28 Treg Expander and remain
Foxp3+.
• Treg cells can be used in any downstream applications (e.g., suppression assays)

Methodology
The isolation procedure includes the following steps:
• Negative isolation of untouched human CD4+ T-cells from PBMC.
• Positive isolation of CD25+ T-cells from the untouched CD4+ T-cell subset.
• Removal of beads from the CD4+CD25+ Treg cells with DETACHaBEAD™

Starting sample
PBMC

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Cell Type T Cells (Regulatory)
Content And Storage Each Kit Contains:
• 2 mL Antibody Mix Human CD4
• 10 mL Depletion MyOne™ Dynabeads™
• 5 mL Dynabeads™ CD25
• 2 mL DETACHaBEAD™.

These reagents are sufficient to process ∼1 x 109 cells. Store components at 2°C to 8°C.
Product Type Cell Isolation Kit
Isolation Technology Depletion, then positive isolation
No. of Cells Processes ∼1 x 109 cells total
Output Viability >95%
Product Line DETACHaBEAD, DYNAL, Dynabeads, FlowComp
Purity or Quality Grade Research Grade
Quantity 2 x 5 mL
Sample Type PBMC
Shipping Condition Room Temperature
Starting Material Cell No. 1 x 108 PBMCs per isolation
Target Species Human
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.