Learn More
Invitrogen™ DES™-Blasticidin Support Kit
Description
Includes
Contains 20μg of pCoBlast, the Calcium Phosphate Transfection Kit, frozen S2 cells, GIBCO Schneider's Drosophila Medium, and 50mg of Blasticidin.
- Contains pCoBlast selection vector for rapid generation of stable S2 cell lines with Blasticidin
- When this kit is paired with pMT/V5-His, pMT/V5-His-TOPO™, pMT-DEST48, pMT/BiP/V5-His, or pAc5.1/V5-His, vector and DES™-Blasticidin Support Kit form complete system for stable expression in Drosophila S2 cells
Constitutive Protein Expression in Insect Cells, Inducible Protein Expression in Insect Cells, Insect Expression, Protein Expression, Proteins, Expression, Isolation and Analysis
Specifications
Specifications
| Product Type | Blasticidin Support Kit |
| Content And Storage | The DES™-Blasticidin Support Kit contains 20 μg of pCoBlast, the Calcium Phosphate Transfection Kit, frozen S2 cells, GIBCO™ Schneider's Drosophila Medium, and 50 mg of Blasticidin. Store S2 cells in liquid nitrogen. Store medium at +4°C. Store all other reagents at -20°C. All components are guaranteed for 6 months when properly stored. |
| Quantity | 1 kit |
| Vector | pCoBlast |
| Product Line | DES |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.