missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cloned AMV First-Strand cDNA Synthesis Kit
Click to view available options
:
100 reactions
Specifications
Specifications
| Content And Storage | 100μL cloned AMV RT (15U/μL), 500μL 5X cDNA synthesis buffer, 2 x 250μL 0.1M DTT, 100μL 10mM dNTP mix, 25μL RnaseOUT (40U/μL), 25μL oligo(dT)20 (50μM), 250μL random hexamers (50ng/μL), 1.25mL DEPC-treated water, product profile sheet. Store at -70deg.C |
| Form | Frozen |
| Quantity | 100 reactions (375 units) |
| Sensitivity | Low |
| For Use With (Application) | cDNA Synthesis, RT-PCR |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction