missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cloned AMV First-Strand cDNA Synthesis Kit
Specifications
Specifications
| Content And Storage | 25μL cloned AMV RT (15U/μL), 500μL 5X cDNA synthesis buffer, 250μL 0.1M DTT, 100μL 10mM dNTP mix, 25μL RnaseOUT (40U/μL), 25μL oligo(dT)20 (50μM), 50μL random hexamers (50ng/μL), 1.25mL DEPC-treated water, product profile sheet. Store at -70deg.C |
| Form | Liquid |
| Format | Tube |
| Quantity | 25 reactions (375 units) |
| Sensitivity | Low |
| For Use With (Application) | cDNA Synthesis, RT-PCR |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?