missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ChargeSwitch™ Wash Buffer W12
Click to view available options
Quantity:
1 L
Unit Size:
Each
Description
Wash buffer used in all ChargeSwitch kits including the ChargeSwitch magnetic beads and ChargeSwitch-Pro column kits.
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Buffer | Wash Buffer |
| Content And Storage | Store at room temperature. |
| Product Line | ChargeSwitch |
| Product Type | Wash Buffer W12 |
| Quantity | 1 L |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction