Learn More
Invitrogen™ ChargeSwitch™-Pro Plasmid Miniprep Kit
The ChargeSwitch™-Pro Plasmid Miniprep Kit generates high-quality plasmid DNA without the use of harsh or potentially interfering reagents, for better success in downstream applications.
£25.26 - £556.00
Specifications
Column Type | Spin Column |
---|---|
Content And Storage | Store at room temperature. |
Format | Kit |
Isolation Technology | ChargeSwitch Chemistry |
Sample Type | Bacterial Culture |
Includes: Miniprep Columns with Collection Tubes, Recovery Tubes, Resuspension Buffer, RNase A, Lysis Buffer, Precipitation Buffer, Wash Buffers 1 and 2, and Elution Buffer
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
10797483
|
Invitrogen™
CS30010 |
10 Preps |
£25.26
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Currently Unavailable: We have no information when this product will be available
View alternative products
|
10717843
|
Invitrogen™
CS30250 |
250 Preps |
£556.00
250 preps |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10707493
|
Invitrogen™
CS30050 |
50 Preps |
£176.00
50 preps |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The ChargeSwitch™-Pro Plasmid Miniprep Kit generates high-quality plasmid DNA without the use of harsh or potentially interfering reagents, for better success in downstream applications.
• 100% ethanol-free, guanidinium-free, organics-free, aqueous protocol for a safer, more streamlined method
• Cleaner purified product for improved performance in downstream applications
• Ready-to-use, complete kit for a fast, hassle-free start
• Ideal for purifying plasmid DNA from 1-5 mL overnight bacterial cultures, yielding up to 20 μg of plasmid DNA
Order Info
Shipping Condition: Room Temperature
Specifications
Spin Column | |
Kit | |
Bacterial Culture | |
Not High-throughput Compatible (Manual) | |
ChargeSwitch™ | |
Room Temperature |
Store at room temperature. | |
ChargeSwitch Chemistry | |
Plasmid DNA | |
< 100 μg (Small-Scale) Plasmid DNA | |
Plasmid MiniPrep Kit | |
Plasmid DNA |
For Research Use Only. Not for use in diagnostic procedures.