Learn More
Invitrogen™ ChargeSwitch™-Pro PCR Clean-Up Kit
The ChargeSwitch™-Pro PCR Clean-up Kit purifies high-quality DNA fragments without the use of ethanol or harsh salts known to interfere with enzymatic reactions.
Brand: Invitrogen™ CS32010
Code : 50
Additional Details : Weight : 0.22000kg
Description
The ChargeSwitch™-Pro PCR Clean-up Kit purifies high-quality DNA fragments without the use of ethanol or harsh salts known to interfere with enzymatic reactions.
• 100% ethanol-free, guanidinium-free, organic solvent-free protocols
• Improved downstream performance
• Complete kit for fast, hassle-free setup
• 20% fewer handling steps than most PCR clean-up protocols
• Purification of up to 500 μL of PCR product in one column
Specifications
The ChargeSwitch™-Pro PCR Clean-up Kit contains columns, collection tubes, recovery tubes, binding buffer, wash buffer, and elution buffer. |
|
Fewer resources used and less waste, Less hazardous | |
ChargeSwitch™ | |
10 Preps | |
DNA from Enzymatic Reactions (e.g. PCR) | |
ChargeSwitch™ Chemistry | |
PCR Products |
Kit | |
Not High-throughput Compatible (Manual) | |
PCR Clean-Up Kit | |
Room Temperature | |
Next-Generation Sequencing (NGS), Cloning, Standard PCR | |
10 Preps |
For Research Use Only. Not for use in diagnostic procedures.