missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ChargeSwitch™ Plasmid Yeast Mini Kit
GREENER_CHOICE

Product Code. 10308732
Click to view available options
Quantity:
50 Preps
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10308732

Brand: Invitrogen™ CS10203

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

  • ChargeSwitch Magnetic Beads (25 mg/mL in 10 mM MES, pH 5.0, 10 mM NaCl, 0.1% Tween 20)
  • RNase A (5 mg/mL in 10 mM Tris-HCl, pH 8.5, 10 mM EDTA)
  • ChargeSwitch Resuspension Buffer (R4)
  • ChargeSwitch Lysis Buffer (L9)
  • ChargeSwitch Precipitation Buffer (N5)
  • ChargeSwitch Wash Buffer (W11)
  • ChargeSwitch Wash Buffer (W12)
  • ChargeSwitch Elution Buffer (E5; 10 mM Tris-HCl, pH 8.5)

The ChargeSwitch Plasmid Yeast Mini Kit provides an efficient method for purifying high-quality plasmid DNA from 1 mL of Saccharomyces cerevisiae grown in selective minimal media.

The ChargeSwitch Plasmid Yeast Mini Kit provides an efficient method for purifying high-quality plasmid DNA from 1 mL of Saccharomyces cerevisiae grown in selective minimal media. Data generated from transformation and PCR experiments using ChargeSwitch purified DNA shows improved performance versus DNA purified by other methods.

  • Sample size 1 mL S. cerevisiae culture
  • Typical yield up to 25 ng
  • Increased transformation efficiency
  • Reagents that avoid PCR inhibition
  • Purifies DNA with improved downstream performance
  • Includes all necessary reagents for number of preps Listed and requires use of magnetic accessory

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage

All components of the ChargeSwitch™ Plasmid Yeast Mini Kit are shipped at room temperature. Upon receipt, store components as follows: Mix RNase A in Resuspension Buffer (R4) and store RNase A in Resuspension Buffer (R4) at 4°C. Store Precipitation Buffer (N5) at 4°C to improve performance. Store the remaining kit components at room temperature. All components are guaranteed stable for 6 months when stored properly.

Components:
• ChargeSwitch™ Magnetic Beads (25 mg/mL in 10 mM MES, pH 5.0, 10 mM NaCl, 0.1% Tween 20)
• RNase A (5 mg/mL in 10 mM Tris-HCl, pH 8.5, 10 mM EDTA)
• ChargeSwitch™ Resuspension Buffer (R4)
• ChargeSwitch™ Lysis Buffer (L9)
• ChargeSwitch™ Precipitation Buffer (N5)
• ChargeSwitch™ Wash Buffer (W11)
• ChargeSwitch™ Wash Buffer (W12)
• ChargeSwitch™ Elution Buffer (E5; 10 mM Tris-HCl, pH 8.5)

Format Kit
Isolation Technology ChargeSwitch Chemistry
Scale Mini
Sample Type Yeast
Final Product Type Plasmid DNA
For Use With (Application) PCR, Cloning, Southern Blotting, Sequencing, Transformation
Green Features Less hazardous, Fewer resources used and less waste
High-throughput Compatibility Not High-throughput Compatible (Manual)
No. of Reactions 50 Preps
Prep Scale < 100 μg (Small-Scale) Plasmid DNA
Product Line ChargeSwitch
Product Type Plasmid Yeast Mini Kit
Quantity 50 Preps
Shipping Condition Room Temperature
Target Plasmid DNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.