missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ChargeSwitch™ Plasmid ER Mini Kit

Product Code. 10653923
Change view
Click to view available options
Quantity:
50 Preps
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10653923 50 Preps Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10653923 Supplier Invitrogen™ Supplier No. CS10100

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

  • ChargeSwitch Magnetic Beads (25 mg/mL) in 10 mM MES, pH 5.0, 10 mM NaCl, 0.1% Tween 20
  • RNase A (5 mg/mL in 10 mM Tris-HCl, pH 8.5, 10 mM EDTA)
  • Resuspension Buffer (R4; 10 mM Tris-HCl, pH 8.5, 10 mM EDTA)
  • ChargeSwitch Lysis Buffer (L9)
  • ChargeSwitch Precipitation Buffer (N5)
  • ChargeSwitch Wash Buffer (W11)
  • ChargeSwitch Wash Buffer (W12)
  • ChargeSwitch Elution Buffer (E5; 10 mM Tris-HCl, pH 8.5)
  • Endotoxin Reduction Reagent (D1; ETRR)

Requires

Magnetic rack

ChargeSwitch™ Plasmid ER Mini Kit uses magnetic bead-based technology to allow for rapid and efficient purification of plasmid DNA.

ChargeSwitch™ Plasmid ER Mini Kit uses magnetic bead-based technology to allow for rapid and efficient purification of plasmid DNA from 1-5 mL of fresh, overnight bacterial cultures without the use of chaotropic salts, organic solvents, or ethanol, to yield up to 20 μg of transfection-grade plasmid DNA in a total time of about 10 minutes after sample preparation.

  • Optimized for higher-purity DNA extraction
  • Improved performance with accompanying magnetic rack
  • Cleaner purified plasmid for optimal downstream performance
  • High quality, high yield plasmid DNA
  • Compatible with MagnaRack™ magnetic rack
  • Reliable performance in variety of downstream applications

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage

All components of the ChargeSwitch™ Plasmid ER (Endotoxin Reduced) Mini Kit are shipped at room temperature. Upon receipt, store components as follows: Store Precipitation Buffer (N5) at 4°C. Mix RNase A in Resuspension Buffer (R4) and store RNase A in Resuspension Buffer (R4) at 4°C. Store the remaining components at room temperature. All components are guaranteed stable for 6 months when stored properly.

Components:
• ChargeSwitch™ Magnetic Beads (25 mg/mL) in 10 mM MES, pH 5.0, 10 mM NaCl, 0.1% Tween 20
• RNase A (5 mg/mL in 10 mM Tris-HCl, pH 8.5, 10 mM EDTA)
• Resuspension Buffer (R4; 10 mM Tris-HCl, pH 8.5, 10 mM EDTA)
• ChargeSwitch™ Lysis Buffer (L9)
• ChargeSwitch™ Precipitation Buffer (N5)
• ChargeSwitch™ Wash Buffer (W11)
• ChargeSwitch™ Wash Buffer (W12)
• ChargeSwitch™ Elution Buffer (E5; 10 mM Tris-HCl, pH 8.5)
• Endotoxin Reduction Reagent (D1; ETRR)

Format Kit
Isolation Technology ChargeSwitch Chemistry
Scale Mini
Sample Type Bacterial Culture
Final Product Type Plasmid DNA
For Use With (Application) Next-Generation Sequencing, Transfection, Cloning, Sequencing, Transformation, Nucleic Acid Labeling, PCR, In Vitro Transcription
High-throughput Compatibility Not High-throughput Compatible (Manual)
Prep Scale < 100 μg (Small-Scale) Plasmid DNA
Product Line ChargeSwitch
Product Type Plasmid ER Mini Kit
Quantity 50 Preps
Shipping Condition Room Temperature
Target Plasmid DNA
Test Time 10 min.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.