missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ChargeSwitch™ NoSpin Plasmid Micro Kit

Product Code. 10756083
Click to view available options
Quantity:
96 Preps
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10756083

Brand: Invitrogen™ CS10201

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

ChargeSwitch NoSpin Kits provide centrifuge-free microprep plasmid purification, ideal for automation.

  • Sample size 0.5–1 mL overnight culture; Typical yield up to 5 μg
  • Provides centrifuge-free purification, ideal for automation
  • Avoids introduction of enzymatic contaminants for better results
  • ChargeSwitch NoSpin Kits provide centrifuge-free microprep plasmid purification, ideal for automation
  • Novel NoSpin reagent causes bacterial cell flocculation and allows cells to be pelleted without the need for automation bottlenecks such as centrifugation
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage

All components of the ChargeSwitch™ NoSpin Plasmid Kits are shipped at room temperature. Upon receipt, store as follows: Store ChargeSwitch™ Precipitation Buffer (N5) at 4°C. Mix the RNase I in ChargeSwitch™ Resuspension Buffer (R4) and store the resulting solution at 4°C. Store all other components at room temperature. All components are guaranteed stable for 6 months if stored properly.

Components:
• ChargeSwitch™ MagnaClear™ Beads
• ChargeSwitch™ Magnetic Beads
• ChargeSwitch™ Resuspension Buffer (R4; 10 mM Tris-HCl, pH 8.5, 10 mM EDTA)
• ChargeSwitch™ Lysis Buffer (L9)
• ChargeSwitch™ Precipitation Buffer (N5)
• ChargeSwitch™ Wash Buffer (W11)
• ChargeSwitch™ Wash Buffer (W12)
• ChargeSwitch™ Elution Buffer (E5; 10 mM Tris-HCl, pH 8.5)
• RNase A (5 mg/mL in 10 mM Tris-HCl, pH 8.5, 10 mM EDTA)

Format Kit
Isolation Technology ChargeSwitch Chemistry
Sample Type Bacterial Culture
Final Product Type Plasmid DNA
For Use With (Application) Next-Generation Sequencing, Transfection, Cloning, Sequencing, Transformation, Nucleic Acid Labeling, PCR, In Vitro Transcription
High-throughput Compatibility High-throughput Compatible (KingFisher Flex)
Prep Scale < 100 μg (Small-Scale) Plasmid DNA
Product Line ChargeSwitch
Product Type NoSpin Plasmid Micro Kit
Quantity 96 Preps
Shipping Condition Room Temperature
Target Plasmid DNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.