missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ChargeSwitch™ Lysis Buffer L13

Click to view available options
Quantity:
800 mL
Unit Size:
Each
Description
- ChargeSwitch Forensic DNA Purification Kit is magnetic bead method designed to accommodate all forensic crime scene sample types for STR profiling
- Scalable for single-tube use or for automated extraction
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Buffer | Lysis Buffer |
| Content And Storage | All components of the ChargeSwitch™ Forensic DNA Purification Kits are shipped at room temperature. Upon receipt, store the Proteinase K at 4°C. Store all other components at room temperature. All components are guaranteed stable for 6 months, if stored properly. |
| Format | Kit |
| For Use With (Application) | DNA Extraction |
| Green Features | Fewer resources used and less waste, Less hazardous |
| Product Line | ChargeSwitch |
| Product Type | Lysis Buffer L13 |
| Quantity | 800 mL |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction