missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ChargeSwitch™ gDNA Plant Kit
GREENER_CHOICE

Product Code. p-4926407
Click to view available options
No. of Reactions:
96 Preps
960 Preps
Sample Type:
Fungi, Plants
Leaf, Seeds
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10695273

Brand: Invitrogen™ CS18000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

ChargeSwitch Magnetic Beads (25 mg/mL in 10 mM MES, pH 5.0, 10 mM NaCl, 0.1% Tween 20)
ChargeSwitch Lysis Buffer (L18)
ChargeSwitch Precipitation Buffer (N5)
ChargeSwitch Wash Buffer (W12)
ChargeSwitch Elution Buffer (E6; 10 mM Tris-HCl, pH 8.5 with 1 mM EDTA)
RNase A (5 mg/mL in 10 mM Tris-HCl, pH 8.5, 10 mM EDTA)
10% SDS in deionized water
10% Detergent (D1)

The ChargeSwitch™ gDNA Plant Kit uses magnetic bead-based technology to allow rapid and efficient isolation of genomic DNA from fungal and plant samples, including leaves, tissues and seeds, in less than 15 minutes after lysate preparation.

The ChargeSwitch™ gDNA Plant Kit uses magnetic bead-based technology to allow rapid and efficient isolation of genomic DNA from fungal and plant samples, including leaves, tissues and seeds, in less than 15 minutes after lysate preparation.

• Optimized for higher-purity DNA extraction
• Compatible with high-throughput protocols
• Ideal for a variety of downstream applications
• Less hazardous, less waste

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage

All components of the ChargeSwitch™ gDNA Plant Kit are shipped at room temperature. Upon receipt, store RNase A at 4°C. Store all other kit components at room temperature.

Components:
• ChargeSwitch™ Magnetic Beads (25 mg/mL in 10 mM MES, pH 5.0, 10 mM NaCl, 0.1% Tween 20)
• ChargeSwitch™ Lysis Buffer (L18)
• ChargeSwitch™ Precipitation Buffer (N5)
• ChargeSwitch™ Wash Buffer (W12)
• ChargeSwitch™ Elution Buffer (E6; 10 mM Tris-HCl, pH 8.5 with 1 mM EDTA)
• RNase A (5 mg/mL in 10 mM Tris-HCl, pH 8.5, 10 mM EDTA)
• 10% SDS in deionized water
• 10% Detergent (D1)

Format Kit
Isolation Technology ChargeSwitch Chemistry
Sample Type Fungi, Plants
For Use With (Application) PCR, Southern Blotting, Sequencing, Cloning
Green Features Fewer resources used and less waste, Less hazardous
High-throughput Compatibility High-throughput Compatible
No. of Reactions 96 Preps
Product Line ChargeSwitch
Product Type gDNA Plant Kit
Quantity 96 Preps
Shipping Condition Room Temperature
Target Genomic DNA
Test Time 15 min.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.