Learn More
Invitrogen™ ChargeSwitch™ gDNA Plant Kit

Description
Includes
ChargeSwitch Magnetic Beads (25 mg/mL in 10 mM MES, pH 5.0, 10 mM NaCl, 0.1% Tween 20)
ChargeSwitch Lysis Buffer (L18)
ChargeSwitch Precipitation Buffer (N5)
ChargeSwitch Wash Buffer (W12)
ChargeSwitch Elution Buffer (E6; 10 mM Tris-HCl, pH 8.5 with 1 mM EDTA)
RNase A (5 mg/mL in 10 mM Tris-HCl, pH 8.5, 10 mM EDTA)
10% SDS in deionized water
10% Detergent (D1)
The ChargeSwitch™ gDNA Plant Kit uses magnetic bead-based technology to allow rapid and efficient isolation of genomic DNA from fungal and plant samples, including leaves, tissues and seeds, in less than 15 minutes after lysate preparation.
• Optimized for higher-purity DNA extraction
• Compatible with high-throughput protocols
• Ideal for a variety of downstream applications
• Less hazardous, less waste
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Content And Storage | All components of the ChargeSwitch™ gDNA Plant Kit are shipped at room temperature. Upon receipt, store RNase A at 4°C. Store all other kit components at room temperature. |
| Format | Kit |
| Isolation Technology | ChargeSwitch Chemistry |
| Sample Type | Leaf, Seeds |
| For Use With (Application) | PCR, Southern Blotting, Sequencing, Cloning |
| High-throughput Compatibility | High-throughput Compatible |
| No. of Reactions | 960 Preps |
| Product Line | ChargeSwitch |
| Product Type | gDNA Plant Kit |
| Quantity | 960 Preps |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.