missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ChargeSwitch™ gDNA Micro Tissue Kit

Product Code. 10523133
Click to view available options
Quantity:
50 Preps
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10523133

Brand: Invitrogen™ CS11203

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Requires

Use of a magnetic accessory.

Sample size mouse ear clips of 3-5 mg tissue with a typical yield up to 5 μg.

  • Sample size mouse ear clips of 3-5 mg tissue; Typical yield up to 5 μg
  • Ultra-sensitive purification from laser microdissected samples
  • Flexibility of an overnight or rapid protocol
  • Reliable PCR performance from a range of micro-tissue samples
  • ChargeSwitch gDNA Micro Tissue Kit provides high levels of performance required to purify high quality DNA from the smallest laser microdissected tissue samples and mouse ear clips
  • Avoids introduction of enzyme inhibitors, which can often limit downstream applications when using very low levels of DNA

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in refrigerator (2–8°C).
Format Kit
Isolation Technology ChargeSwitch Chemistry
Sample Type Tissue
For Use With (Application) PCR, Southern Blotting, Sequencing, Cloning
High-throughput Compatibility Not High-throughput Compatible (Manual)
No. of Reactions 50 Preps
Product Line ChargeSwitch
Product Type gDNA Micro Tissue Kit
Quantity 50 Preps
Shipping Condition Room Temperature
Target Genomic DNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.