missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Champion™ pET161 Gateway™ Expression Kit with Lumio™ Technology

Product Code. 11568716
Change view
Click to view available options
Quantity:
6 μg
Unit Size:
6µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11568716 6 μg 6µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 11568716 Supplier Invitrogen™ Supplier No. 12583043

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

All destination vectors are provided lyophilized and supercoiled.

Requires

Additional materials required, available separately: Gateway™ entry clone, Gateway™ LR Clonase™ enzyme mix, and reaction buffer.

Destination vectors have attR sites for recombination with any attL-flanked fragment, regardless of whether it is entry clone or Ultimate™ RF Clone

Bacterial Expression, Cloning, Enzyme and Protein Activity Assays, Gateway Cloning, Lumio Detection, Protein Expression, Proteins, Expression, Isolation and Analysis, Reporter Assays

TRUSTED_SUSTAINABILITY

Specifications

Constitutive or Inducible System Inducible
Inducing Agent IPTG
Promoter T7
Product Type Expression Kit
Selection Agent (Eukaryotic) None
Content And Storage All destination vectors are provided lyophilized and supercoiled.
Antibiotic Resistance Bacterial Ampicillin (AmpR)
Bacterial or Yeast Strain BL21 Star™(DE3), TOP10
Detection Location In-Gel Detection
Expression System E. coli
Expression Mechanism Cell-Based Expression
Protein Tag His Tag (6x), Lumio
Cloning Method Gateway
Quantity 6 μg
Vector pET
Product Line Champion, Gateway, Lumio
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.