missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cell Suspension Buffer (G1)

Product Code. 12579059
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
This item is not returnable. View return policy

Product Code. 12579059

Brand: Invitrogen™ 12056016

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

500mL bottle

A component in the PureLink 96 Plasmid Purification System

  • The Cell Suspension Buffer (G1) offers a unique approach to high throughput plasmid DNA isolation in the PureLink 96 Plasmid Purification System

Automated Nucleic Acid Purification, Automated Plasmid DNA Purification, DNA & RNA Purification & Analysis, DNA Extraction, Miniprep, Plasmid DNA Purification

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Buffer Suspension Buffer
Quantity 500 mL

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.