Learn More
Invitrogen™ BLOCK-iT™ Lentiviral RNAi Zeo Gateway™ Vector Kit
Product Code. 10798453
Change view
Click to view available options
Quantity:
20 Reactions
Unit Size:
Each
Product Code. Quantity unitSize
10798453 20 Reactions Each
1 options
For Research Use Only. All usage must comply with product instructions.
Product Code. 10798453 Supplier Invitrogen™ Supplier No. V48820
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
For Research Use Only. All usage must comply with product instructions.
Includes
The BLOCK-iT™ Lentiviral RNAi Zeo Gateway™ Vector Kit contains the pLenti4/BLOCK-iT™-DEST vector, pLenti4-GW/H1/TO-lamin shRNA control vector, and One Shot™ Stbl3™ Chemically Competent E.Coli.
For lentiviral delivery and genomic integration of DNA coding for shRNA
The BLOCK-iT Lentiviral RNAi Zeo Gateway Vector Kit contains the pLenti4/BLOCK-iT-DEST expression vector which enables lentiviral delivery and genomic integration of DNA coding for shRNA. Once expressed, the shRNA is processed by cellular machinery and initiates target-specific RNAi.
  • Gateway Technology for efficient recombination of the RNAi cassette from the BLOCK-iT inducible pENTR™/H1/TO vector
  • All required components for efficient lentiviral packaging, delivery, and integration of the shRNA
  • Zeocin selection marker for fast selection of clonal cell lines containing the RNAi cassette
Lentiviral shRNA and miRNA, RNAi, RNAi, Epigenetics and Non-Coding RNA Research, Virus-Based RNAi
TRUSTED_SUSTAINABILITY
Specifications
Product Type RNAi Expression Vector Kit
Content And Storage The BLOCK-iT™ Lentiviral RNAi Zeo Gateway™ Vector Kit contains the pLenti4/BLOCK-iT™-DEST vector, pLenti4-GW/H1/TO-lamin shRNA control vector, and One Shot™ Stbl3™ Chemically Competent E.Coli. Store the pLenti4/BLOCK-iT™-DEST, pLenti6/TR vector, Blasticidin, ViraPower™ Lentiviral packaging mix, and the control vector at -20°C. Store the One Shot Stbl3™ Competent Cells 293 FT cell, and LR Clonase™ enzyme mix at -80°C. Store Lipofectamine™ 2000 at +4°C. All reagents are guaranteed stable for 6 months when properly stored.
Cloning Method Gateway™
Constitutive or Inducible System Inducible
Delivery Type Lentiviral
RNAi Type shRNA
Selection Agent (Eukaryotic) Zeocin™
Vector pLenti
Promoter H1/TO
Format Kit
Product Line BLOCK-iT, Gateway
Quantity 20 Reactions