Learn More
Invitrogen™ BLOCK-iT™ Lentiviral Pol II miR RNAi Expression System
Product Code. 17154529
Change view
Click to view available options
Quantity:
20 Reactions
Unit Size:
20 reactions
Product Code. Quantity Unit Size
17154529 20 Reactions 20 reactions
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 17154529 Supplier Invitrogen™ Supplier No. K493700
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Combines Invitrogen's BLOCK-iT Pol II miR RNAi and ViraPower™ Lentiviral technologies to facilitate creation of a replication-incompetent lentivirus that delivers a microRNA (miRNA) sequence of interest to dividing or non-dividing mammalian cells for RNA interference (RNAi) analysis
Broadens the potential RNAi applications beyond those of other traditional retroviral systems (Naldini, 1998). The BLOCK-iT Lentiviral Pol II miR RNAi Expression System includes:
o A BLOCK-iT Pol II miR RNAi Expression Vector Kit for production of an expression clone containing a double-stranded oligonucleotide encoding a pre-miRNA sequence for expression in mammalian cells. The BLOCK-iT Pol II miR RNAi Expression Vector (pcDNA™6.2-GW/miR) provides a rapid and efficient way to clone ds oligo duplexes encoding a desired miRNA target sequence into a vector containing a Pol II promoter (CMV) for use in RNAi analysis. The BLOCK-iT Pol II miR RNAi Expression Vector is specifically designed to allow expression of miRNA sequences and contain specific miR flanking sequences that allow proper processing of the miRNA.
o The pDONR™221 vector to transfer the premiRNA expression cassette into the lentiviral expression plasmid using Gateway™ Technology. The Gateway Technology is a universal cloning method that takes advantage of the site-specific recombination properties of bacteriophage lambda (Landy, 1989) to provide a rapid and highly efficient way to move your DNA sequence of interest into multiple vector systems.
o A pLenti6/V5-DEST destination vector into which the pre-miRNA cassette from the expression clone is transferred using Gateway Technology. This expression plasmid contains elements that allow packaging of the construct into virions and the Blasticidin resistance marker for selection of stably transduced cell lines.
o Gateway BP and LR Clonase™ II Enzyme Mixes that facilitate the transfer of the pre-miRNA expression cassette from the expression vector into the
pLenti6/V5-DEST destination vector.
o Components of the ViraPower Lentiviral System for production of a replication-incompetent lentivirus that stably expresses the miRNA of interest in both dividing and non-dividing mammalian cells. The ViraPower Lentiviral Technology facilitates highly efficient, in vitro or in vivo delivery of a target gene or RNA to dividing and non-dividing mammalian cells using a replication-incompetent lentivirus. Based on the lentikat™ system developed by Cell Genesys (Dull et al., 1998), the ViraPower Lentiviral Technology possesses features which enhance its biosafety while allowing high-level expression in a wider range of cell types than traditional retroviral systems.
Cloning, Gateway Cloning, Lentiviral shRNA and miRNA, RNAi, RNAi, Epigenetics and Non-Coding RNA Research, Virus-Based RNAi
TRUSTED_SUSTAINABILITY
Especificaciones
Product Type RNAi Expression Vector Kit
Content And Storage The BLOCK-iT™ Lentiviral Pol II miR RNAi Expression System contains the BLOCK-iT™ Pol II miR RNAi Expression Vector Kit which contains two boxes. The cloning box contains linearized pcDNA™6.2-GW/miR, 10X annealing buffer, T4 DNA Ligase, 5X DNA Ligation buffer, lacZ control oligo, lacZ control plasmid, negative control plasmid, DNase/RNase-free water, and forward and reverse sequencing primers. Store the vectors, buffers, control oligo and plasmids, water, and sequencing primers at -20°C. The One Shot™ box contains transformation reagents including twenty-one 50-μl aliquots of One Shot™ TOP10 Chemically Competent E. coli, S.O.C. medium, and a pUC19 supercoiled control plasmid. Store these transformation reagents -80°C. All reagents are guaranteed stable for 6 months when properly stored.
The BLOCK-iT™ Lentiviral Pol II miR RNAi Expression System also includes all the necessary reagents for Gateway™ cloning into the lentiviral expression system, including: pLenti6/V5-DEST Vector, pLenti6-V5/GW-lacZ control, pDONR™221, ViraPower™ Lentiviral Bsd Support Kit (ViraPower™ Packaging Mix, Lipofectamine™ 2000 transfection reagent, and Blasticidin), 293FT Cell Line, One Shot™ Stbl3™ Chemically Competent E. coli, One Shot™ TOP10 Chemically Competent E. coli, and Gateway™ LR and BP Clonase™ II Enzyme Mixes. Store the Lipofectamine™ 2000 reagent at +4°C (do not freeze). Store the vectors, ViraPower™ Packaging Mix, Blasticidin, and both Clonase™ II Enzyme Mixes at -20°C. Store the competent cells at -80°C. Store the 293FT Cell Line in liquid nitrogen. All reagents are guaranteed stable for 6 months when properly stored.
Cloning Method Gateway™
Constitutive or Inducible System Constitutive
Delivery Type Lentiviral
RNAi Type miRNA
Selection Agent (Eukaryotic) Blasticidin
Vector pLenti
Promoter CMV
Product Line BLOCK-iT, Gateway
Quantity 20 Reactions
Mostrar más Mostrar menos
For Research Use Only. Not for use in diagnostic procedures.
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.