missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BioPrime™ Total FFPE Genomic Labeling System

Product Code. 10647995
Change view
Click to view available options
Quantity:
30 Reactions
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10647995 30 Reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10647995 Supplier Invitrogen™ Supplier No. A10965011
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Alexa Fluor™ 3 FFPE 10X nucleotide mix and Alexa Fluor™ 5 FFPE 10X Nucleotide Mix, Exo-Klenow fragment (40U/μL), 5mM EDTA, 2.5X Random Primers solution, control DNA (Salmon Sperm), and DEPC-treated water, 10X Nucleotide Mixes, Purelink™ Genomic DNA purification columns and purification buffers.

Complete genomic DNA labeling kit designed for use in array comparative genomic hybridization applications (aCGH) using formalin-fixed paraffin-embedded (FFPE) samples

Previous protocols have required large amounts of very high quality DNA, which is often difficult to obtain from FFPE material. With a random prime amplification (RPA) method, using labeling mixes specifically formulated for FFPE samples, BioPrime™ Total FFPE Genomic Labeling System generates high yields of labeled DNA which is most representative of the starting sample. Increased call accuracy is achieved with higher yields of labeled material on the array, giving you more accurate calls.

This complete, all-inclusive kit is optimized to work across a wider range of sample input material with no need for pre-amplification. Highly concentrated exo-Klenow fragment of DNA polymerase I in a component limited reaction allow for consistent, robust DNA yields of genomic FFPE DNA input. BioPrime™ Total for Agilent™ aCGH is supplied with Purelink™ purification which allows for low elution, eliminating the need for complicated volume-reduction steps. Purelink™ purification also reduces background due to dye bled found in other purification systems An optimized dye labeled nucleotide mix with new dNTP linkers, novel, application-specific Alexa Fluor™ 3 and 5 dyes, and improved buffer chemistry reduce labeling variation and increase signal to noise on arrays. Excitation and emission spectra of Alexa Fluor™ 3 and 5 dyes are suited for conventional two color scanners with no need to change settings. Complete kits are batch tested to assure quality in performance from lot to lot.

  • Improved call rate with higher signal to noise ratios from Alexa Fluor™ 3 and 5 dyes
  • Easy cleanup with PureLink™ purification eliminates the need for complicated volume-reduction steps
  • Less channel bias due to enhanced reaction formulation for FFPE samples
  • Lower requirements for input material using enzymatic RPA method

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains Alexa Fluor™ 3 FFPE 10X nucleotide mix and Alexa Fluor™ 5 FFPE 10X Nucleotide Mix, Exo-Klenow fragment (40 U⁄μl), 5 mM EDTA, 2.5X Random Primers solution, control DNA (Salmon Sperm), and DEPC-treated water. Store at -80°C for long term storage. The 10X Nucleotide Mixes may be stored at +4°C for up to 4 weeks and should be protected from light. Also contains Purelink™ Genomic DNA purification columns and purification buffers. Store at room temperature. All components are guaranteed for 6 months when properly stored.
Detection Method Fluorescence
Label or Dye Alexa Fluor 3, Alexa Fluor 5
Final Product Type Probes (Labeled DNA)
Sample Type Tissue (FFPE)
Labeling Target Genomic DNA
Excitation/Emission 650/670, 555/565
Includes Label or Dye Yes
Labeling Method Direct Labeling
Product Line BioPrime
Product Type Genomic Labeling System
Quantity 30 Reactions
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.