Learn More
Invitrogen™ BenchPro™ 2100 Plasmid Processing Station
High-throughput compatibility, automated protocols
Brand: Invitrogen™ MC1001
Code : NEW
Additional Details : Weight : 0.00450kg
Includes: BenchPro 2100 Plasmid Processing Station, AC/DC Switching Power Adapter, 2 Waste Trays, Piercing device, Quick-disconnect house air fitting including 12 ft. tubing, Quick Reference Card (QRC)
Description
The BenchPro 2100 Plasmid Purification System is an innovative, automated instrument that provides a completely walk-away solution for high quality purification of large-scale plasmid DNA.
- Innovative, straight-from-culture automation
- Superior yield and purity
- Minimize hands-on work and human errors
- Reagent type: Cell Lysis Buffer
- Loading volume: 100-125mL bacterial culture
- Reaction time: 90 minutes
- Capacity: Up to two preps per run
- 13W x 20.8D x 11.7 in.H
Innovative, Straight-From-Culture Automation Going straight from culture to purified plasmid, the BenchPro 2100 simplifies the standard, 22-step process of manual plasmid purification down a few easy set-up steps that can be performed typically in less than five minutes. Assemble the Reagent Tray and Cell Liner on the Waste Tray, and add a 125mL E. coli culture to the Cell Liner. Load the BenchPro 2100 Plasmid Purification Card into an instrument slot, and run the purification protocol. Unparalleled Yield and Purity The BenchPro 2100 Plasmid Purification Card at the core of the instrument is a disposable, fluidics card that contains a novel anion-exchange membrane that provides the same high-quality plasmid DNA as our manual PureLink™ HiPure kits provide. Each plasmid preparation can be trusted for use in many applications—mammalian cell transfection, automated and manual sequencing, PCR amplification, cloning, transformation, and in vitro transcription. The BenchPro 2100 system is able to reproducibly purify a range of plasmid sizes, from 2.7 to 20kb. Plasmid yields from the BenchPro 2100 are unmatched by those achieved by alternative methods. Minimizes Variation and User Error The BenchPro 2100 Plasmid Purification System is designed to provide consistency and reliability by using prefilled and sealed reagents. All required buffers in appropriate volumes are supplied in a sealed Reagent Tray, and a disposable Cell Liner (for the addition of culture) with lid is included. The BenchPro 2100 Plasmid Purification System accurately performs all steps of the purification process to help deliver consistent yield, eliminating the tedious and repetitive liquid-handling steps associated with the manual process of plasmid purification. During the automated purification protocol, predefined amounts of each reagent are sequentially pumped into the card, and incubation steps performed according to the protocol. Using a series of capture and purification membranes, E. coli cells are harvested and lysed, and the lysate clarified. By minimizing the number of hands-on steps, the BenchPro 2100 Plasmid Purification System enhances experimental reproducibility and data consistency run after run.
Automated Nucleic Acid Purification, Automated Plasmid DNA Purification, DNA & RNA Purification & Analysis, DNA Extraction, Maxiprep, Plasmid DNA Purification
Order Info
Shipping Condition: Room temperature
Specifications
Automated maxiprep purification of plasmid DNA | |
BenchPro 2100 Plasmid Processing Station, AC/DC Switching Power Adapter, 2 Waste Trays, Piercing device, Quick-disconnect house air fitting including 12 ft. tubing, Quick Reference Card (QRC), Instruction Manual |
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.