missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BaculoDirect™ N-Term Transfection Kit
Click to view available options
:
5 transfections
Description
Includes
Five 0.3μg vials of BaculoDirect Linear DNA, 125μL of Cellfectin transfection reagent, and ganciclovir.
- Engineered to include attR sites for quick and efficient recombination with Gateway entry clone
- Gene of interest is recombined from entry clone into BaculoDirect Linear DNA using a simple, one-hour LR reaction
- Resulting reaction mix contains recombinant baculovirus carrying gene of interest and is used directly to transfect insect cells
- Need for transforming bacteria and isolating large bacmid or co-transfection of a transfer vector and linear baculovirus DNA into insect cells is eliminated for reduced hands-on time
- Purified baculovirus can be isolated in less than one week
- N-terminal V5-His tag for detection with Anti-V5 Antibody and purification with nickel-chelating resin (BaculoDirect N-Term Linear DNA)
- TEV protease cleavage site for removal of V5-His tag following purification (BaculoDirectN-Term Linear DNA)
- C-terminal V5-His tag for detection with Invitrogens Anti-V5 Antibody and purification with nickel-chelating resin (BaculoDirect C-Term Linear DNA)
- Expression can be detected as early as first baculovirus infection
- Gateway Linear DNA: Designed for rapid cloning with a Gateway entry clone and subsequent expression in Sf9 or Sf21 insect cells
- Strong polyhedrin promoter for high-level expression
- R sites for efficient recombination with any attL-flanked Gateway entry vector
- TK gene for negative selection using ganciclovir
- N-terminal V5-His tag for detection with Invitrogens Anti-V5 Antibody and purification with nickel-chelating resin (BaculoDirect N-Term Linear DNA)
- TEV protease cleavage site for removal of the V5-His tag following purification (BaculoDirect N-Term Linear DNA)
- C-terminal V5-His tag for detection with Invitrogens Anti-V5 Antibody and purification with nickel-chelating resin (BaculoDirect C-Term Linear DNA)
Baculovirus Production and Titering/Insect Expression, Protein Expression, Proteins, Expression, Isolation and Analysis
Specifications
Specifications
| Promoter | Polyhedrin |
| Product Type | Transfection Kit |
| Content And Storage | The BaculoDirect™ transfection kits include five 0.3 μg vials of BaculoDirect Linear DNA, 125 μL Cellfectin transfection reagent, and ganciclovir. Store the BaculoDirect Linear DNA and Cellfectin at 4°C. All components are guaranteed stable for 6 months if stored properly. |
| Cleavage | TEV Protease Recognition Site |
| Protein Tag | His Tag (6x), V5 Epitope Tag |
| Cloning Method | Gateway |
| Vector | BaculoDirect Vectors |
| Product Line | BaculoDirect™, Gateway™ |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction