Learn More
Gibco™ Bac-to-Bac™ HT Vector Kit
Description
Includes
The Bac-to-Bac HT Vector Kit includes 10μg of pFastBac HT vector and pFastBac HT-CAT control vector.
- Strong polyhedrin promoter for protein expression
- Three reading frames for simplified cloning
- N-terminal 6 x His tag for simple purification of recombinant fusion proteins
- TEV protease cleavage site for removal of histidine tag following protein purification
Baculovirus Production and Titering, Insect Expression, Protein Expression, Proteins, Expression, Isolation and Analysis
Specifications
Specifications
| Promoter | Polyhedrin |
| Product Type | Vector Kit |
| Content And Storage | The Bac-to-Bac™ HT Vector Kit includes 10 μg of pFastBac™ HT vector and pFastBac HT-CAT control vector. Store the vectors at -20°C. All components are guaranteed stable for 6 months when properly stored. |
| Cleavage | TEV Protease Recognition Site |
| Protein Tag | His Tag (6x) |
| Cloning Method | Restriction Enzyme/MCS |
| Quantity | 1 kit |
| Vector | pFastBac |
| Product Line | Bac-to-Bac |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?