Learn More
Invitrogen™ Anchored Oligo(dT)20 Primer
Product Code. 11666131
Click to view available options
:
50 μg
Product Code. 11666131
Brand: Invitrogen™ 12577011
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Anchored Oligo(dT)20 Primer is a primer mixture consisting of a string of 20 deoxythymidylic acid residues followed by dV (dG, dA, or dC) and then by dN (dA, dT, dG, or dC).
Anchored Oligo(dT)20 Primer is a primer mixture consisting of a string of 20 deoxythymidylic acid residues followed by dV (dG, dA, or dC) and then by dN (dA, dT, dG, or dC). Due to the variable end-sequence, the primer is anchored to the 5´-end of the poly(A) tail of mRNA and prevents priming within the poly(A) tail.
Anchored Oligo(dT)20 Primer is recommended for use in cDNA labeling protocols and in RT-PCR. It is commonly used for first-strand cDNA systhesis.
Quality control: This product is qualified in a first-strand cDNA synthesis reaction
Order Info
Shipping Conditions: Dry Ice